3-Amino-5-bromobenzonitrile 99%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
5-Amino-3-bromobenzonitrile
OR63032-10G
OR63032-1G
OR63032-5G
77410-810EA
205.8
USD
77410-810
77410-812
77410-814
3-Amino-5-bromobenzonitrile 99%
3-Amino-5-bromobenzonitrile
Formula:
C₇H₅BrN₂ MW: 197.0300 g/mol Melting Pt: 124…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD11109874 CAS Number: 49674-16-0 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...