6-Bromopyridine-2-carbonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR2804-25G OR2804-5G
77408-020EA 263.88 USD
77408-020 77408-022
6-Bromopyridine-2-carbonitrile 98%
6-Bromopyridine-2-carbonitrile
Formula: C₆H₃BrN₂
Melting Pt: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00234142
CAS Number: 122918-25-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR