4-Bromo-1H-indazole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Bromoindazole

OR12759-10G OR12759-25G OR12759-5G
77404-420EA 28.52 USD
77404-420 77404-422 77404-424
4-Bromo-1H-indazole
4-Bromo-1H-indazole
Formula: C₇H₅BrN₂
MW: 197.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05664001
CAS Number: 186407-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR