3(2H)-Pyridazinone 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,3-dihydropyridazin-3-one, Pyridazin-3(2H)-one

OR2215-100G OR2215-25G OR2215-5G
77406-804EA 207.84 USD
77406-804 77406-810 77406-820
3(2H)-Pyridazinone 95%
3(2H)-Pyridazinone
Formula: C₄H₄N₂O
MW: 96.0874 g/mol
Boiling Pt: 101 °C/1.5 mmHg
Melting Pt: 98…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00099605
CAS Number: 504-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR