3-Methoxybenzenesulfonamide

Supplier: APOLLO SCIENTIFIC
Warning

OR2656-1G OR2656-250MG OR2656-5G
77407-806EA 103.92 USD
77407-806 77407-812 77407-816
3-Methoxybenzenesulfonamide
3-Methoxybenzenesulphonamide
Formula: C₇H₉NO₃S
Melting Pt: 131…132 °C
Storage Temperature: Ambient
MDL Number: MFCD08704579
CAS Number: 58734-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR