4-(2-Thiazolylcarbamoyl)phenylboronic acid 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR1192-1G
OR1192-50MG
OR1192-5G
77404-060EA
227.2
USD
77404-060
77404-062
77404-064
4-(2-Thiazolylcarbamoyl)phenylboronic acid 98%
4-(2-Thiazolylcarbamoyl)phenylboronic acid
Formula:
C₁₀H₉BN₂O₃S Melting Pt: 206…210 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659877 CAS Number: 850568-26-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...