2,2-Difluoro-2-(fluorosulfonyl)acetic acid 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC9975-100G PC9975-25G
77418-906EA 158.94 USD
77418-906 77418-908
2,2-Difluoro-2-(fluorosulfonyl)acetic acid 98%
2,2-Difluoro-2-(fluorosulphonyl)acetic acid

Methyl and trimethylsilyl esters also available.

Formula: C₂HF₃O₄S
Boiling Pt: 153 °C
Density: 1.723 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01320807
CAS Number: 1717-59-5
UN: 2927
ADR: 6.1+8,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR