5-(2-Nitrophenyl)oxazole

Supplier: APOLLO SCIENTIFIC
Warning

OR24051-1G OR24051-500MG
77407-410EA 256.74 USD
77407-410 77407-412
5-(2-Nitrophenyl)oxazole
5-(2-Nitrophenyl)oxazole
Formula: C₉H₆N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00085146
CAS Number: 89808-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR