4-Iodo-5-methylisoxazole 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR14344-1G OR14344-250MG
77404-920EA 227.2 USD
77404-920 77404-922
4-Iodo-5-methylisoxazole 97%
4-Iodo-5-methylisoxazole
Formula: C₄H₄INO
Melting Pt: 27…28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06411541
CAS Number: 7064-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR