5-Amino-7-bromo-1H-indazole 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 7-Bromo-1H-indazol-5-amine

OR110843-1G OR110843-250MG
77403-504EA 28.46 USD
77403-504 77403-506
5-Amino-7-bromo-1H-indazole 98%
5-Amino-7-bromo-1H-indazole
Formula: C₇H₆BrN₃
MW: 212.0500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08275114
CAS Number: 953411-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR