3-Iodo-5-methyl-1H-indazole

Supplier: APOLLO SCIENTIFIC
Warning

OR18425-1G OR18425-250MG
77406-336EA 59.1 USD
77406-336 77406-338
3-Iodo-5-methyl-1H-indazole
3-Iodo-5-methyl-1H-indazole
Formula: C₈H₇IN₂
MW: 258.0600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781621
CAS Number: 885518-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR