Tri(2-furyl)phosphine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Tri(furan-2-yl)phosphine, Tri-2-furylphosphine

OR10819-1G OR10819-250MG
77402-936EA 34.64 USD
77402-936 77402-938
Tri(2-furyl)phosphine
Tri(2-furyl)phosphine
Formula: C₁₂H₉O₃P
MW: 232.17 g/mol
Boiling Pt: 136 °C/4 mmHg
Melting Pt: 59…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00151857
CAS Number: 5518-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR