6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR21501-25G OR21501-5G
77406-694EA 330.1 USD
77406-694 77406-696
6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid
6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid
Formula: C₅H₆N₂O₃
MW: 142.12 g/mol
Melting Pt: 199…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00052030
CAS Number: 27372-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR