Methyl-2-(4-bromophenyl)imidazo[1,2-a]pyridine-8-carboxylate

Supplier: APOLLO SCIENTIFIC

OR15102-1G OR15102-250MG
77405-290EA 410.58 USD
77405-290 77405-292
Methyl-2-(4-bromophenyl)imidazo[1,2-a]pyridine-8-carboxylate
Methyl 2-(4-bromophenyl)imidazo[1,2-a]pyridine-8-carboxylate
Formula: C₁₅H₁₁BrN₂O₂
Melting Pt: 134…136 °C
Storage Temperature: Ambient
MDL Number: MFCD08444011
CAS Number: 133427-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR