3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile

Supplier: APOLLO SCIENTIFIC

Synonyms: 3-Bromo-6-cyanoimidazo[1,2-a]pyridine

OR110193-1G OR110193-5G
77403-122EA 131.42 USD
77403-122 77403-124
3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile
3-Bromoimidazo[1,2-a]pyridine-6-carbonitrile
Formula: C₈H₄BrN₃
MW: 222.05 g/mol
Melting Pt: 207…209 °C
Storage Temperature: Ambient
MDL Number: MFCD06659617
CAS Number: 885950-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR