4-(Ethylsulphonyl)phenylboronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (4-(Ethylsulphonyl)phenyl)boronic acid, 4-(Ethylsulphonyl)benzeneboronic acid

OR10479-1G OR10479-500MG
77402-734EA 124.3 USD
77402-734 77402-736
4-(Ethylsulphonyl)phenylboronic acid 97%
4-(Ethylsulphonyl)phenylboronic acid
Formula: C₈H₁₁BO₄S
Melting Pt: 135…139 °C
Storage Temperature: Ambient
MDL Number: MFCD02093076
CAS Number: 352530-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR