Methyl-6-bromoimidazo[1,2-a]pyridine-3-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR110059-1G OR110059-250MG OR110059-5G
77403-042EA 227.2 USD
77403-042 77403-044 77403-046
Methyl-6-bromoimidazo[1,2-a]pyridine-3-carboxylate
Methyl 6-bromoimidazo[1,2-a]pyridine-3-carboxylate
Formula: C₉H₇BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD21364589
CAS Number: 1359656-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR