Ethyl-6-bromoimidazo[1,2-a]pyridine-3-carboxylate 95%

Supplier: APOLLO SCIENTIFIC

Synonyms: Ethyl 6-bromoimidazo[1,2-a]pyridine-3-carboxylate

OR300507-1G
77408-672EA 61 USD
77408-672
Ethyl-6-bromoimidazo[1,2-a]pyridine-3-carboxylate 95%
Ethyl 6-bromoimidazo[1,2-a]pyridine-3-carboxylate
Formula: C₁₀H₉BrN₂O₂
MW: 269.1000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10698068
CAS Number: 372198-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR