Methyl-3,4-dimethylbenzoate

Supplier: APOLLO SCIENTIFIC
Warning

OR25704-1G OR25704-25G OR25704-5G
77407-628EA 30 USD
77407-628 77407-630 77407-632
Methyl-3,4-dimethylbenzoate
Methyl 3,4-dimethylbenzoate
Formula: C₁₀H₁₂O₂
Melting Pt: 35…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00219861
CAS Number: 38404-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR