4-(Methoxycarboxy)-1-naphthaleneboronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR11435-1G OR11435-250MG
77403-914EA 981.12 USD
77403-914 77403-916
4-(Methoxycarboxy)-1-naphthaleneboronic acid 98%
4-(Methoxycarboxy)-1-naphthaleneboronic acid
Formula: C₁₂H₁₁BO₄
Melting Pt: 170…173 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09475824
CAS Number: 957034-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR