(4-Cyanophenyl)methanesulfonyl chloride, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

OR43515-1G OR43515-250MG
77409-702EA 118.8 USD
77409-702 77409-704
(4-Cyanophenyl)methanesulfonyl chloride, Technical Grade
(4-Cyanophenyl)methanesulfonyl chloride
Formula: C₈H₆ClNO₂S
Melting Pt: 99.5…101.5 °C
Storage Temperature: Ambient
MDL Number: MFCD07690517
CAS Number: 56105-99-8
UN: 2923
ADR: 8+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR