4-Nitro-3-(trifluoromethyl)benzenesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

PC6567-1G PC6567-5G
77417-436EA 96.78 USD
77417-436 77417-438
4-Nitro-3-(trifluoromethyl)benzenesulfonyl chloride
4-Nitro-3-(trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₃ClF₃NO₄S
Melting Pt: 61…67 °C
Storage Temperature: Ambient
MDL Number: MFCD03094387
CAS Number: 39234-83-8
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR