3,4-Difluorosalicylaldehyde 95%

Supplier: APOLLO SCIENTIFIC

Synonyms: 3,4-Difluoro-2-hydroxybenzaldehyde

PC201259-1G PC201259-250MG PC201259-5G
77413-474EA 74.38 USD
77413-474 77413-476 77413-478
3,4-Difluorosalicylaldehyde 95%
3,4-Difluorosalicylaldehyde
Formula: C₇H₄F₂O₂
MW: 158.1023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08275283
CAS Number: 502762-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR