(1-Ethoxycarbonylethylidene)triphenylphosphorane 95%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
(Carbethoxyethylidene)triphenylphosphorane, Ethyl 2-(triphenylphosphoranylidene)propanoate
OR10643-100G
OR10643-25G
77402-848EA
74.38
USD
77402-848
77402-850
(1-Ethoxycarbonylethylidene)triphenylphosphorane 95%
(1-Ethoxycarbonylethylidene)triphenylphosphorane
Formula:
C₂₃H₂₃O₂P MW: 362.41 g/mol Melting Pt: 157…159 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009160 CAS Number: 5717-37-3 EINECS: 227-215-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...