(R)-(+)-1-Boc-3-aminopyrrolidine 95%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
(R)-1-Boc-3-aminopyrrolidine, (3R)-(+)-1-Boc-3-aminopyrrolidine, (3R)-(+)-3-Amino-1-Boc-pyrrolidine, (R)-tert-Butyl 3-aminopyrrolidine-1-carboxylate, (3R)-(+)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine, (3R)-(+)-3-Amino-1-(tert-butoxycarbonyl)pyrrolidine
OR4635-25G
OR4635-5G
77409-930EA
68.26
USD
77409-930
77409-932
(R)-(+)-1-Boc-3-aminopyrrolidine 95%
(R)-(+)-1-Boc-3-aminopyrrolidine
Formula:
C₉H₁₈N₂O₂ Boiling Pt: 243…244 °C Density: 1.098 g/cm³ (25 °C) Flash Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD03419272 CAS Number: 147081-49-0 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...