(Iodomethyl)cyclopropane 95%

Supplier: APOLLO SCIENTIFIC

OR111605-10G OR111605-1G OR111605-5G
77403-826EA 496.16 USD
77403-826 77403-828 77403-830
(Iodomethyl)cyclopropane 95%
(Iodomethyl)cyclopropane
Formula: C₄H₇I
MW: 182.00 g/mol
Boiling Pt: 53…54 °C
Storage Temperature: Ambient
MDL Number: MFCD06410734
CAS Number: 33574-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR