Dysprosium(III) sulfate octahydrate 99,9%

Supplier: APOLLO SCIENTIFIC
Warning

IN1669-100G IN1669-25G
77401-424EA 389.58 USD
77401-424 77401-426
Dysprosium(III) sulfate octahydrate 99,9%
Dysprosium (III) sulphate octahydrate
Formula: Dy₂O₁₂S₃·8H₂O
Storage Temperature: Ambient
CAS Number: 10031-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR