Perfluoroacetophenone

Supplier: APOLLO SCIENTIFIC
Warning

PC7644-1G PC7644-25G
77418-076EA 63.16 USD
77418-076 77418-078 77418-080
Perfluoroacetophenone
Perfluoroacetophenone
Formula: C₈F₈O
MW: 264.072 g/mol
Boiling Pt: 130…131 °C
Density: 1.609 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00216596
CAS Number: 652-22-2
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR