2,2,3,4,4,4-Hexafluorobutyl methacrylate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,2,3,4,4,4-Hexafluorobutyl Methacrylate

PC4657-100G PC4657-25G PC4657-5G
77415-656EA 227.2 USD
77415-656 77415-658 77415-660
2,2,3,4,4,4-Hexafluorobutyl methacrylate 98%
2,2,3,4,4,4-Hexafluorobutyl methacrylate

Stabilised; Refrigeration will extend shelf-life.

Formula: C₈H₈F₆O₂
MW: 250.14 g/mol
Boiling Pt: 158 °C
Density: 1.348 g/cm³ (25 °C)
Flash Pt: 57 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042311
CAS Number: 36405-47-7
UN: 3272
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR