6-Nitro-o-anisidine
Supplier: APOLLO SCIENTIFIC
Synonyms:
6-Nitro-o-anisidine, 2-Amino-3-nitroanisole, 2-Methoxy-6-nitrophenylamine, 2-Methoxy-6-nitroaniline
OR300658-1G
OR300658-5G
77408-680EA
21.26
USD
77408-680
77408-682
6-Nitro-o-anisidine
6-Nitro-o-anisidine
Formula:
C₇H₈N₂O₃ MW: 168.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01930197 CAS Number: 16554-45-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...