3-Hydroxyisonicotinic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR12100-1G OR12100-25G OR12100-5G
77404-174EA 21.26 USD
77404-174 77404-176 77404-178
3-Hydroxyisonicotinic acid
3-Hydroxyisonicotinic acid
Formula: C₆H₅NO₃
Melting Pt: 315…319 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00234165
CAS Number: 10128-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR