Ethyl pentafluoropropionate 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC3250-1G PC3250-25G PC3250-5G
77414-604EA 21.26 USD
77414-604 77414-606 77414-608
Ethyl pentafluoropropionate 98%
Ethyl pentafluoropropionate

Undergoes Wittig reaction to give 1-perfluoroalkylenol ethers, which are useful precursors to other fluorine containing molecules: JOC., 57, 3807 (1992).

Formula: C₅H₅F₅O₂
Boiling Pt: 73…76 °C
Density: 1.299 g/cm³ (25 °C)
Flash Pt: 1 °C
Storage Temperature: Ambient
MDL Number: MFCD00000431
CAS Number: 426-65-3
EINECS: 207-043-6
UN: 3272
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR