4,4-Difluorocyclohexanol 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC51054-5G PC51054-1G
77416-434EA 241.46 USD
77416-434 77416-432
4,4-Difluorocyclohexanol 95%
4,4-Difluorocyclohexanol
Formula: C₆H₁₀F₂O
MW: 136.1398 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10000566
CAS Number: 22419-35-8
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR