3-Fluoro-5-(trifluoromethoxy)aniline

Supplier: APOLLO SCIENTIFIC
Warning

PC302847-1G PC302847-250MG PC302847-5G
77414-288EA 389.18 USD
77414-288 77414-290 77414-292
3-Fluoro-5-(trifluoromethoxy)aniline
3-Fluoro-5-(trifluoromethoxy)aniline
Formula: C₇H₅F₄NO
Storage Temperature: Ambient
MDL Number: MFCD18783229
CAS Number: 1352999-96-2
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR