2-Amino-3-fluoro-5-(trifluoromethyl)pyridine 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC5457-5G
PC5457-1G
PC5457-25G
77416-754EA
58.9
USD
77416-754
77416-750
77416-752
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine 97%
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine
Formula:
C₆H₄F₄N₂ Melting Pt: 64…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD10699117 CAS Number: 852062-17-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...