Difluoroacetic anhydride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC3791-100G PC3791-25G PC3791-5G
77414-892EA 144.68 USD
77414-892 77414-894 77414-896
Difluoroacetic anhydride 97%
Difluoroacetic anhydride
Formula: C₄H₂F₄O₃
Boiling Pt: 125…127 °C
Density: 1.4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02093315
CAS Number: 401-67-2
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR