N-Methyl 4-boronobenzenesulfonamide 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Methylsulphamoylbenzeneboronic acid, (4-(N-Methylsulphamoyl)phenyl)boronic acid

OR1324-1G OR1324-250MG
77404-600EA 140.6 USD
77404-600 77404-602
N-Methyl 4-boronobenzenesulfonamide 98%
N-Methyl 4-boronobenzenesulphonamide
Formula: C₇H₁₀BNO₄S
MW: 215.03 g/mol
Melting Pt: 136…140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659864
CAS Number: 226396-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR