4-(1-Pyrrolidinyl)pyridine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Pyrrolidinopyridine

OR1965-100G OR1965-25G OR1965-5G
77406-506EA 124.3 USD
77406-506 77406-508 77406-510
4-(1-Pyrrolidinyl)pyridine
4-(1-Pyrrolidinyl)pyridine
Formula: C₉H₁₂N₂
MW: 148.21 g/mol
Melting Pt: 54…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00006415
CAS Number: 2456-81-7
UN: 2923
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR