4-Chlorophenyl chloroformate
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR13039-100G
OR13039-25G
OR13039-5G
77404-494EA
406.5
USD
77404-494
77404-496
77404-498
4-Chlorophenyl chloroformate
4-Chlorophenyl chloroformate
Formula:
C₇H₄Cl₂O₂ Boiling Pt: 100…102 °C/12 mmHg Density: 1.365 g/cm³ (25 °C) Flash Pt: 103 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000638 CAS Number: 7693-45-0 UN: 3277 ADR: 6.1+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...