2,2,2-Trifluoroethyl trichloromethanesulfonate 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC7377-1G
PC7377-5G
77417-846EA
48.9
USD
77417-846
77417-848
2,2,2-Trifluoroethyl trichloromethanesulfonate 97%
2,2,2-Trifluoroethyl trichloromethanesulphonate
Formula:
C₃H₂Cl₃F₃O₃S Boiling Pt: 84…86 °C/20 mmHg Melting Pt: –70 °C Density: 1.714 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00042400 CAS Number: 23199-56-6 EINECS: 245-484-6 UN: 2927 ADR: 6.1+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...