2-Bromo-2'-fluoro-4'-methoxyacetophenone 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Bromo-1-(2-fluoro-4-methoxyphenyl)ethanone, 2-Fluoro-4-methoxyphenacyl bromide

PC1154-1G PC1154-250MG PC1154-5G
77413-066EA 69.28 USD
77413-066 77413-068 77413-070
2-Bromo-2'-fluoro-4'-methoxyacetophenone 97%
2-Bromo-2'-fluoro-4'-methoxyacetophenone
Formula: C₉H₈BrFO₂
Melting Pt: 63…66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03730100
CAS Number: 157014-35-2
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR