3-Chloro-4-(trifluoromethoxy)benzyl bromide 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC0181-1G
PC0181-25G
PC0181-5G
77412-434EA
36.68
USD
77412-434
77412-436
77412-438
3-Chloro-4-(trifluoromethoxy)benzyl bromide 98%
3-Chloro-4-(trifluoromethoxy)benzyl bromide
Formula:
C₈H₅BrClF₃O Storage Temperature: Ambient |
MDL Number:
MFCD01631560 CAS Number: 261763-18-2 UN: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...