α,3-Dibromo-4-fluorotoluene 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Bromo-4-fluorobenzyl bromide, 2-Bromo-4-(bromomethyl)-1-fluorobenzene

PC9668-5G PC9668-100G PC9668-25G
77418-714EA 34.64 USD
77418-714 77418-710 77418-712
α,3-Dibromo-4-fluorotoluene 98%
α,3-Dibromo-4-fluorotoluene
Formula: C₇H₅Br₂F
Boiling Pt: 130 °C/0.5 mmHg
Melting Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD09037367
CAS Number: 78239-71-1
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR