2-Amino-5-(trifluoromethyl)benzonitrile

Supplier: APOLLO SCIENTIFIC
Danger

PC3059-1G PC3059-5G
77414-440EA 21.26 USD
77414-440 77414-442
2-Amino-5-(trifluoromethyl)benzonitrile
2-Amino-5-(trifluoromethyl)benzonitrile
Formula: C₈H₅F₃N₂
Boiling Pt: 95…115 °C/0.4 mmHg
Melting Pt: 72…74 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03407465
CAS Number: 6526-08-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR