3-Chloro-2,4-difluorobenzylamine 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (3-Chloro-2,4-difluorophenyl)methanamine

PC302828-1G PC302828-5G
77414-282EA 103.92 USD
77414-282 77414-284
3-Chloro-2,4-difluorobenzylamine 98%
3-Chloro-2,4-difluorobenzylamine
Formula: C₇H₆ClF₂N
MW: 177.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660335
CAS Number: 886761-65-5
UN: 2735
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR