2,3-Dichloro-6-fluorobenzylamine 97%

Supplier: APOLLO SCIENTIFIC

PC302006-1G PC302006-5G
77414-094EA 101.88 USD
77414-094 77414-096
2,3-Dichloro-6-fluorobenzylamine 97%
2,3-Dichloro-6-fluorobenzylamine
Formula: C₇H₆Cl₂FN
Storage Temperature: Ambient
MDL Number: MFCD06660174
CAS Number: 886501-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR