4-Aminotetrahydropyran hydrochloride 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR10343-1G OR10343-25G OR10343-5G
77402-610EA 21.26 USD
77402-610 77402-612 77402-614
4-Aminotetrahydropyran hydrochloride 97%
4-Aminotetrahydropyran hydrochloride
Formula: C₅H₁₂ClNO
Melting Pt: 230…250 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00100881
CAS Number: 33024-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR