2-Bromo-2'-fluoro-3'-(trifluoromethyl)acetophenone

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Fluoro-3-(trifluoromethyl)phenacyl bromide

PC8405-1G PC8405-25G PC8405-5G
77418-406EA 54.52 USD
77418-406 77418-408 77418-410
2-Bromo-2'-fluoro-3'-(trifluoromethyl)acetophenone
2-Bromo-2'-fluoro-3'-(trifluoromethyl)acetophenone
Formula: C₉H₅BrF₄O
Melting Pt: 46.9…47.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094504
CAS Number: 537050-11-6
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR