2-(Methylthio)benzylamine
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR0657-1G
OR0657-250MG
77402-180EA
221.08
USD
77402-180
77402-182
2-(Methylthio)benzylamine
2-(Methylthio)benzylamine
Formula:
C₈H₁₁NS Boiling Pt: 139…141 °C/15 mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00831039 CAS Number: 56004-83-2 UN: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...