2-Bromo-4'-(trifluoromethoxy)acetophenone 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Bromo-1-(4-(trifluoromethoxy)phenyl)ethanone, 4-(Trifluoromethoxy)phenacyl bromide

PC7439KM-100G PC7439KM-25G PC7439KM-5G
77417-944EA 215.98 USD
77417-944 77417-946 77417-948
2-Bromo-4'-(trifluoromethoxy)acetophenone 97%
2-Bromo-4'-(trifluoromethoxy)acetophenone
Formula: C₉H₆BrF₃O₂
MW: 283.04 g/mol
Melting Pt: 48…55 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052333
CAS Number: 103962-10-3
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR